missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO44 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24386
This item is not returnable.
View return policy
Description
FBXO44 Polyclonal specifically detects FBXO44 in Human samples. It is validated for Western Blot.
Specifications
| FBXO44 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 93611 | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| FBXO44 | |
| Reacts with residues 103-119 (EWKVEDLSRDQRKEFPN) of the human FBG3 protein. | |
| 0.1 mg | |
| Primary | |
| High cross reactivity: 94% identical to mouse and rat FBG3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction