missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXO44 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 360.00 €
Specifications
| Antigen | FBXO44 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18159352
|
Novus Biologicals
NBP2-24386SS |
0.025 mg |
198.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18148911
|
Novus Biologicals
NBP2-24386 |
0.1 mg |
360.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FBXO44 Polyclonal specifically detects FBXO44 in Human samples. It is validated for Western Blot.Specifications
| FBXO44 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 93611 | |
| Reacts with residues 103-119 (EWKVEDLSRDQRKEFPN) of the human FBG3 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FBG3 DKFZp781J0852, F-box gene 3, F-box only protein 44, F-box protein 44, F-box protein FBX30, F-box/G-domain protein 3, FBX30FBX6AFbx44, FBX44, Fbxo6a, MGC14140 | |
| FBXO44 | |
| IgG | |
| Affinity Purified | |
| High cross reactivity: 94% identical to mouse and rat FBG3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title