missing translation for 'onlineSavingsMsg'
Learn More

FBXW11 Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™

Product Code. 30513641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30513641 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30513641 Supplier Novus Biologicals Supplier No. NBP338464AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FBXW11 Polyclonal antibody specifically detects FBXW11 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXW11
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias beta-transducin repeat-containing protein 2, BTRC2F-box/WD repeat-containing protein 1B, BTRCP2FBW1B, F-box and WD repeat domain containing 11, F-box and WD-40 domain protein 11, F-box and WD-40 domain protein 1B, F-box protein Fbw1b, F-box/WD repeat-containing protein 11, Fbw11, FBXW1BFbw1b, Homologous to Slimb protein, Hos, KIAA0696F-box and WD repeats protein beta-TrCP2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 443-542 of human FBXW11 (NP_036432.2).,, Sequence:, MEPDSVIEDKTIELMCSVPRSLWLGCANLVESMCALSCLQSMPSVRCLQISNGTSSVIVSRKRPSEGNYQKEKDLCIKYFDQWSE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 23291
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.