missing translation for 'onlineSavingsMsg'
Learn More

FCAR/CD89 Antibody (14A8 (14.1)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Product Code. 18653895 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.02mg
0.2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18653895 0.025 mg 0.02mg
18664904 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18653895 Supplier Novus Biologicals Supplier No. NBP3201610.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FCAR/CD89 Monoclonal antibody specifically detects FCAR/CD89 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCAR/CD89
Applications Flow Cytometry, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 14A8 (14.1)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS
Gene Alias CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Host Species Rabbit
Immunogen The original antibody was generated by immunizing HCO12 trangene containing mice with recombinant soluble CD89 antigen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2204
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.