missing translation for 'onlineSavingsMsg'
Learn More

FcgR4/CD16-2 Antibody (012), Novus Biologicals™

Product Code. p-200058542 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18604246 100 μg 100µL
18621926 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18604246 Supplier Novus Biologicals Supplier No. NBP305890100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FcgR4/CD16-2 Monoclonal antibody specifically detects FcgR4/CD16-2 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcgR4/CD16-2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 012
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:50-1:1000
Formulation 0.2 um filtered solution in PBS
Gene Alias 4833442P21Rik;CD16-2;Fcgam;FcgammaRIV;Fcgr;Fcgr3a;FcgRIV;Fcrl3;Low affinity immunoglobulin gamma Fc region receptor III-A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FcgR4/CD16-2 (Accession#: NP_653142.2; Met 1-Gln 203).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246256
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.