missing translation for 'onlineSavingsMsg'
Learn More
Learn More
fd/M13 bacteriophage Antibody (MM05), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05788-50ul
This item is not returnable.
View return policy
Description
fd/M13 bacteriophage Monoclonal antibody specifically detects fd/M13 bacteriophage in Virus samples. It is validated for ELISA
Specifications
| fd/M13 bacteriophage | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 927333 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| MM05 | |
| ELISA 1:2500-1:10000 | |
| major coat protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with M13 Bacteriophage. | |
| 50 μg | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction