missing translation for 'onlineSavingsMsg'
Learn More
Learn More
fd/M13 bacteriophage Antibody (MM05), Novus Biologicals™
Mouse Monoclonal Antibody
231.00 € - 326.00 €
Specifications
| Antigen | fd/M13 bacteriophage |
|---|---|
| Clone | MM05 |
| Dilution | ELISA 1:2500-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685248
|
Novus Biologicals
NBP3-05788-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622217
|
Novus Biologicals
NBP3-05788-50ul |
50 μg |
231.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
fd/M13 bacteriophage Monoclonal antibody specifically detects fd/M13 bacteriophage in Virus samples. It is validated for ELISASpecifications
| fd/M13 bacteriophage | |
| ELISA 1:2500-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 927333 | |
| IgG1 | |
| Protein A purified |
| MM05 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Virus | |
| major coat protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with M13 Bacteriophage. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title