missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FDPS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30807-100ug
This item is not returnable.
View return policy
Description
FDPS Polyclonal antibody specifically detects FDPS in Nonspecies specific samples. It is validated for ELISA,Immunocytochemistry/ Immunofluorescence
Specifications
| FDPS | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Non-species specific | |
| Purified |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293 | |
| Rabbit immunized with farnesyl cysteine conjugated to KLH. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 2224 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction