missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FDPS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 564.00 €
Specifications
| Antigen | FDPS |
|---|---|
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066987
|
Novus Biologicals™
NBP3-30807-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067241
|
Novus Biologicals™
NBP3-30807-100ug |
100 μg |
564.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FDPS Polyclonal antibody specifically detects FDPS in Nonspecies specific samples. It is validated for ELISA,Immunocytochemistry/ ImmunofluorescenceSpecifications
| FDPS | |
| Polyclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293 | |
| Rabbit immunized with farnesyl cysteine conjugated to KLH. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| Phosphate buffered saline | |
| 2224 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title