missing translation for 'onlineSavingsMsg'
Learn More

Fenclofenac Ethyl Ester, Mikromol™

Product Code. 17980356
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980356 Supplier Mikromol Supplier No. MM3075.02
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fenclofenac Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenclofenac Ethyl Ester
CAS 86308-84-1
Analyte or Component Names Fenclofenac Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H14 Cl2 O3
Certifications/Compliance ISO 9001
Synonym 2-(2,4-Dichlorophenoxy)benzeneacetic acid ethyl ester
InChI Formula InChI=1S/C16H14Cl2O3/c1-2-20-16(19)9-11-5-3-4-6-14(11)21-15-8-7-12(17)10-13(15)18/h3-8,10H,2,9H2,1H3
SMILES CCOC(=O)Cc1ccccc1Oc2ccc(Cl)cc2Cl
IUPAC Name ethyl 2-[2-(2,4-dichlorophenoxy)phenyl]acetate
Molecular Weight (g/mol) 325.19
Quantity 100 mg
Formula Weight 324.032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.