missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30324559
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324559 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324559 Supplier Dr. Ehrenstorfer Supplier No. DREA13486100AL100
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenofibric Acid
CAS 42017-89-0
Analyte or Component Names Fenofibric Acid
Shipping Condition +20°C
Molecular Formula C17 H15 Cl O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Fenofibrate Imp. B (EP), Fenofibrate USP Related Compound B, Fenofibrate USP RC B, 2[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid, Fenofibric Acid, Propanoic acid, 2-[4-(4-chlorobenzoyl)phenoxy]-2-methyl-, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid, 2-[4'-(p-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, 2-[p-(p-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, FNF acid, LF 153, LF 178 acid, NSC 281318, Procetofenic acid, alpha-1081, Fenofibrate Related Compound B, Fenofibrate Impurity B
InChI Formula InChI=1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)O
IUPAC Name 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 318.75
Quantity 1 mL
Formula Weight 318.0659 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.