missing translation for 'onlineSavingsMsg'
Learn More

Fenspiride Hydrochloride, Mikromol™

Product Code. 17953886
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953886 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17953886 Supplier Mikromol Supplier No. MM3773.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fenspiride Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenspiride Hydrochloride
CAS 8-7-5053
Analyte or Component Names Fenspiride Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C15 H20 N2 O2 . Cl H
Certifications/Compliance ISO 17025
Synonym 1-Oxa-3,8-diazaspiro[4.5]decan-2-one, 8-(2-phenylethyl)-, hydrochloride (1:1), 1-Oxa-3,8-diazaspiro[4.5]decan-2-one, 8-(2-phenylethyl)-, monohydrochloride (9CI), 1-Oxa-3,8-diazaspiro[4.5]decan-2-one, 8-phenethyl-, monohydrochloride (8CI), 8-Phenethyl-1-oxa-3,8-diazaspiro[4.5]decan-2-one hydrochloride, Decaspir, Decaspiride, Erespal, Fenspiride hydrochloride, Fluiden, JP 428, NAT 333, NDR 5998A, Pneumorel, Tegencia, Viarespan
InChI Formula InChI=1S/C15H20N2O2.ClH/c18-14-16-12-15(19-14)7-10-17(11-8-15)9-6-13-4-2-1-3-5-13;/h1-5H,6-12H2,(H,16,18);1H
SMILES Cl.O=C1NCC2(CCN(CCc3ccccc3)CC2)O1
IUPAC Name 8-(2-phenylethyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one;hydrochloride
Molecular Weight (g/mol) 296.79
Quantity 250 mg
Formula Weight 296.1292 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.