missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin B Antibody (SR1975), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22629-100ul
This item is not returnable.
View return policy
Description
Fetuin B Monoclonal antibody specifically detects Fetuin B in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Serum
Specifications
| Fetuin B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26998 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| SR1975 | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
| FETUB, fetuin beta;fetuin phosphoprotein, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
| A synthesized peptide derived from human Fetuin B (Uniprot #: Q9UGM5) | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction