missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fetuin B Antibody (SR1975), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 449.00 €
Specifications
| Antigen | Fetuin B |
|---|---|
| Clone | SR1975 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611265
|
Novus Biologicals
NBP3-22629-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604165
|
Novus Biologicals
NBP3-22629-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fetuin B Monoclonal antibody specifically detects Fetuin B in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, SerumSpecifications
| Fetuin B | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 26998 | |
| IgG | |
| Affinity purified |
| SR1975 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| FETUB, fetuin beta;fetuin phosphoprotein, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685 | |
| A synthesized peptide derived from human Fetuin B (Uniprot #: Q9UGM5) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title