missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine EP Impurity E (Terfenadine EP Impurity I), Mikromol™

Product Code. 30050865
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050865 25 mg 25mg
17900766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050865 Supplier Mikromol Supplier No. MM0055.020025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fexofenadine EP Impurity E (Terfenadine EP Impurity I)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azacyclonol
CAS 115-46-8
Analyte or Component Names Azacyclonol
Shipping Condition Room Temperature
Molecular Formula C18 H21 N O
Certifications/Compliance ISO 17025
Synonym Diphenyl(piperidin-4-yl)methanol (Azacyclonol), Fexofenadine Hydrochloride Imp. E (EP), Fexofenadine Imp. E (EP), Terfenadine Imp. I (EP), Diphenyl(piperidin-4-yl)methanol, Azacyclonol, Pipradrol 4-Isomer, Fexofenadine Hydrochloride Impurity E, Terfenadine Impurity I, Fexofenadine Impurity E
SMILES OC(C1CCNCC1)(c2ccccc2)c3ccccc3
Molecular Weight (g/mol) 267.37
Quantity 25 mg
Formula Weight 267.372 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.