Learn More
FGF basic/FGF2/bFGF Rabbit anti-Human, Clone: 13, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89192-100UL
 View more versions of this product
Product Code. 18004876
  • 594.00 € / 100µL
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF basic/FGF2/bFGF
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
13
Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
A synthetic peptide corresponding to the N-terminus of the Human FGF basic/FGF2/bFGF
100 μg
Angiogenesis, Biologically Active Proteins, Cancer
2247
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.