missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Rabbit anti-Human, Clone: 13, Novus Biologicals™
Rabbit Monoclonal Antibody
392.00 € - 594.00 €
Specifications
| Antigen | FGF basic/FGF2/bFGF |
|---|---|
| Clone | 13 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18285517
|
Novus Biologicals
NBP2-89192-50UL |
50 μg |
392.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18004876
|
Novus Biologicals
NBP2-89192-100UL |
100 μg |
594.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FGF basic/FGF2/bFGF | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| A synthetic peptide corresponding to the N-terminus of the Human FGF basic/FGF2/bFGF | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 13 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 0.2 um filtered solution in PBS | |
| 2247 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title