missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FGF basic/FGF2/bFGF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
208.00 € - 2080.00 €
Specifications
| Antigen | FGF basic/FGF2 |
|---|---|
| Clone | 10060 |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797119
![]() |
R&D Systems
MAB233-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773947
![]() |
R&D Systems
MAB233-100 |
100 μg |
517.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762657
![]() |
R&D Systems
MAB233-500 |
500 μg |
2080.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| FGF basic/FGF2 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2247 | |
| E. coli-derived recombinant human FGF basic/FGF2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Recognizes human and bovine FGF basic/FGF2. In capture ELISAs, shows approximately 0.2% cross-reactivity with bovine FGF acidic, and no cross-reactivity with recombinant human (rh) FGF acidic, rhFGF-4, rhFGF-6, or rhFGF-7. |
| 10060 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin | |
| FGF2 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title