Learn More
Invitrogen™ FGFR1OP Recombinant Rabbit Monoclonal Antibody (23GB5705), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553102
 View more versions of this product
Product Code. 30088997
  • 446.00 € / 100µL
Description
This gene encodes a largely hydrophilic protein postulated to be a leucine-rich protein family member. A t(6;8)(q27;p11) chromosomal translocation, fusing this gene and the fibroblast growth factor receptor 1 (FGFR1) gene, has been found in cases of myeloproliferative disorder. The resulting chimeric protein contains the N-terminal leucine-rich region of this encoded protein fused to the catalytic domain of FGFR1. This gene is thought to play an important role in normal proliferation and differentiation of the erythroid lineage. Alternatively spliced transcript variants that encode different proteins have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGFR1OP
Recombinant Monoclonal
Unconjugated
FGFR1OP
4930553O10Rik; centrosomal protein 43; centrosomal protein 43; FGFR1 oncogene partner; CEP43; FGFR1 oncogene partner; FGFR1 oncogene partner 1; FGFR1 oncoprotein partner-like protein; Fgfr1op; fibroblast growth factor receptor 1 oncogene partner; FOP; LOC683722; Unknown (protein for MGC:134294)
Rabbit
Affinity chromatography
RUO
11116, 683722, 75296
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5705
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95684, Q4V7C1, Q66JX5
FGFR1OP
A synthesized peptide derived from human CEP43 (350-399AA).
100 μL
Primary
Human, Mouse, Rat