missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGFR1OP Recombinant Rabbit Monoclonal Antibody (23GB5705), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
186.00 € - 446.00 €
Specifications
| Antigen | FGFR1OP |
|---|---|
| Clone | 23GB5705 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088997
![]() |
Invitrogen™
MA553102 |
100 μL |
446.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089940
|
Invitrogen™
MA553103 |
20 μL |
186.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR1OP Recombinant Monoclonal antibody specifically detects FGFR1OP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| FGFR1OP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O95684, Q4V7C1, Q66JX5 | |
| 11116, 683722, 75296 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5705 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4930553O10Rik; centrosomal protein 43; centrosomal protein 43; FGFR1 oncogene partner; CEP43; FGFR1 oncogene partner; FGFR1 oncogene partner 1; FGFR1 oncoprotein partner-like protein; Fgfr1op; fibroblast growth factor receptor 1 oncogene partner; FOP; LOC683722; Unknown (protein for MGC:134294) | |
| A synthesized peptide derived from human CEP43 (350-399AA). | |
| Primary | |
| -20°C | |
| FGFR1OP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title