missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FHL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30885-100ug
This item is not returnable.
View return policy
Description
FHL2 Polyclonal antibody specifically detects FHL2 in Human samples. It is validated for Western Blot
Specifications
| FHL2 | |
| Polyclonal | |
| Western Blot : 5-2 μg/mL | |
| down-regulated in rhabdomyosarcoma LIM protein, DRALAAG11, FHL-2, four and a half LIM domains 2, four and a half LIM domains protein 2, four and a half LIM-domain protein 2, LIM domain protein DRAL, Skeletal muscle LIM-protein 3, SLIM-3, SLIM3aging-associated gene 11 | |
| Synthetic peptide derived from the human FHL2 protein | |
| 100 μg | |
| Angiogenesis, Cancer | |
| 2274 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction