missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FHL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | FHL2 |
|---|---|
| Dilution | Western Blot : 5-2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067067
|
Novus Biologicals™
NBP3-30885-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066939
|
Novus Biologicals™
NBP3-30885-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FHL2 Polyclonal antibody specifically detects FHL2 in Human samples. It is validated for Western BlotSpecifications
| FHL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer | |
| Phosphate buffered saline | |
| 2274 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| down-regulated in rhabdomyosarcoma LIM protein, DRALAAG11, FHL-2, four and a half LIM domains 2, four and a half LIM domains protein 2, four and a half LIM-domain protein 2, LIM domain protein DRAL, Skeletal muscle LIM-protein 3, SLIM-3, SLIM3aging-associated gene 11 | |
| Synthetic peptide derived from the human FHL2 protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title