Learn More
Fibroblast Antibody (ER-TR7), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NB100-64932AF350
Product Code. 18769870
  • 628.00 € / 125µL
Description
Fibroblast Monoclonal antibody specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibroblast
Monoclonal
Alexa Fluor 350
reticular fibroblast
Isolated C3H thymic stromal cells
0.125 mL
Fibroblast Cell Markers
Mouse
Purified
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
ER-TR7
50mM Sodium Borate
Rat
Protein G purified
RUO
Primary
Store at 4°C in the dark.
IgG2a