missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Surface Protein Antibody (1B10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-1845-0.025ml
This item is not returnable.
View return policy
Description
Fibroblast Surface Protein Monoclonal specifically detects Fibroblast Surface Protein in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Fibroblast Surface Protein | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Complement-dependent Cytotoxicity 1:500 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Flow Cytometry | |
| 1B10 | |
| Unconjugated | |
| The product is provided as the immunoglobulin fraction of mouse ascites fluid in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA. with 15mM Sodium Azide | |
| cultured human thymic fibroblasts | |
| 0.025 mL | |
| Core ESC Like Genes, Fibroblast Cell Markers, Stem Cell Markers | |
| This fibroblast antigen has been demonstrated on human synovial, mammary, foreskin and thymic fibroblasts and in malignant fibrosarcoma tissue. The antigen is both cell membrane and lysosome associated. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction