missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Surface Protein Antibody (1B10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
222.00 €
Specifications
| Antigen | Fibroblast Surface Protein |
|---|---|
| Clone | 1B10 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Complement-dependent Cytotoxicity 1:500 |
| Applications | Flow Cytometry |
Description
Fibroblast Surface Protein Monoclonal specifically detects Fibroblast Surface Protein in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| Fibroblast Surface Protein | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Fibroblast Cell Markers, Stem Cell Markers | |
| The product is provided as the immunoglobulin fraction of mouse ascites fluid in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA. with 15mM Sodium Azide | |
| IgM | |
| Unpurified | |
| This fibroblast antigen has been demonstrated on human synovial, mammary, foreskin and thymic fibroblasts and in malignant fibrosarcoma tissue. The antigen is both cell membrane and lysosome associated. |
| 1B10 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Complement-dependent Cytotoxicity 1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| cultured human thymic fibroblasts | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title