missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FITC Polyclonal antibody specifically detects FITC in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, ELISA
Specifications
Specifications
| Antigen | FITC |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Fluorescein Isothiocyanate |
| Host Species | Sheep |
| Immunogen | Pooled antisera from sheep hyperimmunized with fluorescein isothiocyanate (Isomer I) |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?