missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Fixed Price Planned Maintenance for Incubators

Product Code. 30503413
Change view
Click to view available options
Product Type:
Ovens
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30503413 Ovens Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30503413 Supplier Thermo Scientific Supplier No. FPPMCOV
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fixed-cost planned maintenance service for incubators, environmental chambers, vacuum ovens, and furnaces-designed to ensure consistent operation and reduce unplanned downtime.

  • Fixed-price service coverage with no hidden costs
  • Supports a range of laboratory equipment
    • Incubators
    • Environmental chambers
    • Vacuum ovens
    • Laboratory furnaces
  • Helps maintain optimal performance and temperature control
  • Reduces unexpected downtime and costly repairs
  • Performed by certified service professionals
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Incubators, environmental chambers, vacuum ovens, furnaces
Duration 1 Day
Product Type Ovens
Target Audience Employees/Users
Species All
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.