Learn More
Thermo Scientific Fixed Price Preventative Maintenance for Centrifugal Evaporator
Product Code. 32018035 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Centrifugal Evaporator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
32018035 Centrifugal Evaporator Each
1 options
Product Code. 32018035 Supplier Thermo Scientific Supplier No. FPPMEVAPMVS
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes one scheduled on-site preventative maintenance visit for centrifugal evaporators by a trained IES (Unity Lab Services) engineer.
  • Service includes inspection, functional checks, vacuum performance review, temperature and rotor speed checks where applicable, safety checks, and maintenance documentation within the agreed labor scope
  • PM kits, spare parts, pump service kits, seals, high-cost consumables and repairs are excluded unless separately confirmed and quoted
  • Additional labor, travel or repeat visits caused by customer/site readiness issues may be charged in line with the applicable rate card
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Centrifugal Evaporator
Product Type Preventive Maintenance