missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Mouse, Clone: FLI1/1312, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2313-MSM2-P1
Additional Details : Weight : 0.01000kg
Specifications
FLI1 | |
Nuclear | |
FLI1/1312 | |
Unconjugated | |
NP_001161153 | |
Mouse | |
51kDa | |
Protein A/G Purified | |
RUO | |
2313 | |
IgG |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FLI1 | |
Recombinant fragment (aa 107-261) of human FLI1 protein | |
THP1 and Raji cells. Tonsil, Ewing s sarcoma (EWS) and Angiosarcoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |