missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Mouse, Clone: FLI1/1312, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | FLI1 |
---|---|
Clone | FLI1/1312 |
Concentration | 200μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959997
|
EnquireBio
2313-MSM2-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949997
|
EnquireBio
2313-MSM2-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
FLI1 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
2313 | |
Recombinant fragment (aa 107-261) of human FLI1 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
51kDa |
FLI1/1312 | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001161153 | |
FLI1 | |
IgG | |
Protein A/G Purified | |
Nuclear | |
THP1 and Raji cells. Tonsil, Ewing s sarcoma (EWS) and Angiosarcoma |