missing translation for 'onlineSavingsMsg'
Learn More

Flotillin-1 Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30493498 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30493498 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30493498 Supplier Novus Biologicals Supplier No. NBP337979FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Flotillin-1 Polyclonal antibody specifically detects Flotillin-1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Flotillin-1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias flotillin 1, flotillin-1, integral membrane component of caveolae
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 128-427 of human Flotillin-1 (NP_005794.1).,, Sequence:, HDDQDYLHSLGKARTAQVQKDARIGEAEAKRDAGIREAKAKQEKVSAQYLSEIEMAKAQRDYELKKAAYDIEVNTRRAQADLAYQLQVAKTKQQIEEQRVQVQVVERAQQVAVQEQEIARREKELEARVRKPAEAERYKLERLAEAEKSQLIMQAEAEAASVRMRGEAEAFAIGARARAEAEQMAKKAEAFQLYQEAAQLDMLLEKLPQVAEEISGPLTSANKITLVSSGSGTMGAAKVTGEVLDILTRLPESVERLTGVSISQVNHKPLRTA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10211
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.