missing translation for 'onlineSavingsMsg'
Learn More

Fluazuron 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30324203
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324203 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324203 Supplier Dr. Ehrenstorfer Supplier No. DREA13672000AL1000
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluazuron
CAS 86811-58-7
Analyte or Component Names Fluazuron
Shipping Condition +20°C
Molecular Formula C20 H10 Cl2 F5 N3 O3
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym N-[[[4-Chloro-3-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]amino]carbonyl]-2,6-difluorobenzamide, Acatak, CGA 157419, Fluazuron
InChI Formula InChI=1S/C20H10Cl2F5N3O3/c21-11-5-4-10(29-19(32)30-17(31)16-13(23)2-1-3-14(16)24)7-15(11)33-18-12(22)6-9(8-28-18)20(25,26)27/h1-8H,(H2,29,30,31,32)
SMILES Fc1cccc(F)c1C(=O)NC(=O)Nc2ccc(Cl)c(Oc3ncc(cc3Cl)C(F)(F)F)c2
IUPAC Name N-[[4-chloro-3-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenyl]carbamoyl]-2,6-difluorobenzamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 506.21
Quantity 1 mL
Formula Weight 505.0019 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.