missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole EP Impurity D, Mikromol™

Product Code. 30050461
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050461 25 mg 25mg
30051205 100 mg 100mg
30461132 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050461 Supplier Mikromol Supplier No. MM0463.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flubendazole EP Impurity D
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1H-Benzimidazol-5-yl)(4-fluorophenyl)methanone
CAS 1515807-30-3
Analyte or Component Names (1H-Benzimidazol-5-yl)(4-fluorophenyl)methanone
Shipping Condition Room Temperature
Molecular Formula C14 H9 F N2 O
Certifications/Compliance ISO 17025
Synonym (1H-Benzimidazol-5-yl)(4-fluorophenyl)methanone, Flubendazole Imp. D (EP), Flubendazole Impurity D
InChI Formula InChI=1S/C14H9FN2O/c15-11-4-1-9(2-5-11)14(18)10-3-6-12-13(7-10)17-8-16-12/h1-8H,(H,16,17)
SMILES Fc1ccc(cc1)C(=O)c2ccc3[nH]cnc3c2
IUPAC Name 1H-benzimidazol-5-yl-(4-fluorophenyl)methanone
Molecular Weight (g/mol) 240.23
Quantity 25 mg
Formula Weight 240.0699 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.