missing translation for 'onlineSavingsMsg'
Learn More

Flunarizine Dihydrochloride, LoGiCal

Product Code. 30458668
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458668 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458668 Supplier LoGiCal Supplier No. LGCFOR0175.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Flunarizine dihydrochloride
CAS 30484-77-6
Analyte or Component Names Flunarizine dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C26 H26 F2 N2 . 2 Cl H
Certifications/Compliance ISO 9001
Synonym Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenyl-2-propen-1-yl]-, hydrochloride (1:2), Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)-, dihydrochloride, (E)-, Piperazine, 1-[bis(4-fluorophenyl)methyl]-4-[(2E)-3-phenyl-2-propenyl]-, dihydrochloride (9CI), Piperazine, 1-[bis(p-fluorophenyl)methyl]-4-cinnamyl-, dihydrochloride, (E)- (8CI), (E)-1-[Bis(4-fluorophenyl)methyl]-4-cinnamylpiperazine dihydrochloride, Dinaplex, Flugeral, Flunagen, Flunarizine dihydrochloride, Flunarizine hydrochloride, Flunarl, Fluxarten, Gradient, Issium, Mondus, R 14950
InChI Formula InChI=1S/C26H26F2N2.2ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)30-19-17-29(18-20-30)16-4-7-21-5-2-1-3-6-21;;/h1-15,26H,16-20H2;2*1H/b7-4+;;
Solution Type Neat
SMILES Cl.Cl.Fc1ccc(cc1)C(N2CCN(C\C=C\c3ccccc3)CC2)c4ccc(F)cc4
IUPAC Name 1-[bis(4-fluorophenyl)methyl]-4-[(E)-3-phenylprop-2-enyl]piperazine;dihydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 477.42
Quantity 10 mg
Formula Weight 476.1598 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.