missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flunixin Meglumine EP Impurity A (Niflumic Acid EP Impurity A), Mikromol™
Description
Flunixin Meglumine EP Impurity A (Niflumic Acid EP Impurity A)
Specifications
Specifications
| Chemical Name or Material | 2-Chloronicotinic acid |
| CAS | 2942-59-8 |
| Analyte or Component Names | 2-Chloronicotinic acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H4 Cl N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Chloropyridine-3-carboxylic Acid, Flunixin Meglumine Imp. A (EP), Niflumic Acid Imp. A (EP), Flunixin USP Related Compound A, Flunixin USP RC A, 2-Chloropyridine-3-carboxylic Acid, 2-Chloronicotinic Acid, Flunixin Related Compound A, Flunixin Meglumine Impurity A, Niflumic Acid Impurity A, Flunixin Impurity A |
| InChI Formula | InChI=1S/C6H4ClNO2/c7-5-4(6(9)10)2-1-3-8-5/h1-3H,(H,9,10) |
| SMILES | OC(=O)c1cccnc1Cl |
| IUPAC Name | 2-chloropyridine-3-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?