missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoxetine Hydrochloride, Mikromol™
Description
Fluoxetine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Fluoxetine Hydrochloride |
| CAS | 56296-78-7 |
| Analyte or Component Names | Fluoxetine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 F3 N O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Fluoxetine Hydrochloride, Adofen, Affectine, Alzac 20, Ansilan, Deproxin, Digassim, Erocap, Fluctin, Fluctine, Fludac, Flufran, Flunil, Fluox-Puren, Fluoxac, Fluoxeren, Fluoxetine hydrochloride, Fluoxil, Flutin, Flutine, Fluxen, Fluxetyl, Fluxil, Fontex, Foxetin, LY 110140, Lilly 110140, Lorien, Lovan, Margrilan, Modipran, N-Methyl-3-(4-trifluoromethylphenoxy)-3-phenylpropylamine hydrochloride, Neupax, Nodepe, Nopres, Nuzak, Octozac, Oxedep, Pluzac, Pragmaten, Prizma, Proctin, Prodep, Profluzac, Prozac, Prozac 20, Reconcile, Reneuron, Rowexetina, Salipax, Sanzur, Sarafem, Sinzac, Zactin, Zedprex, Zepax, N-Methyl-gamma-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride (1:1), (+/-)-N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine hydrochloride |
| InChI Formula | InChI=1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H |
| SMILES | Cl.CNCCC(Oc1ccc(cc1)C(F)(F)F)c2ccccc2 |
| IUPAC Name | N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Hittar du en möjlighet till förbättring?