missing translation for 'onlineSavingsMsg'
Learn More

Flurbiprofen Isopropyl Ester, Mikromol™

Product Code. 30050212
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050212 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050212 Supplier Mikromol Supplier No. MM0727.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flurbiprofen Isopropyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flurbiprofen Isopropyl Ester
CAS 252846-89-2
Analyte or Component Names Flurbiprofen Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C18 H19 F O2
Certifications/Compliance ISO 9001
Synonym Flurbiprofen Isopropyl Ester, 2-Fluoro-alpha-methyl-[1,1'-biphenyl]-4-acetic acid 1-methylethyl ester
InChI Formula InChI=1S/C18H19FO2/c1-12(2)21-18(20)13(3)15-9-10-16(17(19)11-15)14-7-5-4-6-8-14/h4-13H,1-3H3
SMILES CC(C)OC(=O)C(C)c1ccc(c(F)c1)c2ccccc2
IUPAC Name propan-2-yl 2-(3-fluoro-4-phenylphenyl)propanoate
Molecular Weight (g/mol) 286.34
Quantity 100 mg
Formula Weight 286.1369 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.