missing translation for 'onlineSavingsMsg'
Learn More

Flutamide, Mikromol™

Product Code. 18042810
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042810 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18042810 Supplier Mikromol Supplier No. MM0161.000250
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flutamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flutamide
CAS 13311-84-7
Analyte or Component Names Flutamide
Shipping Condition Room Temperature
Molecular Formula C11 H11 F3 N2 O3
Certifications/Compliance ISO 17034
Synonym Flutamide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide, 4'-Nitro-3'-trifluoromethylisobutyranilide, 4-Nitro-3-(trifluoromethyl)isobutyranilide, Euflex, Eulexin, Flucinom, N-(Isopropylcarbonyl)-4-nitro-3-trifluoromethylaniline, NSC 147834, NSC 215876, Niftholide, Niftolide, Odyne, Sch 13521, alpha,alpha,alpha-Trifluoro-2-methyl-4'-nitro-m-propionotoluidide
InChI Formula InChI=1S/C11H11F3N2O3/c1-6(2)10(17)15-7-3-4-9(16(18)19)8(5-7)11(12,13)14/h3-6H,1-2H3,(H,15,17)
SMILES CC(C)C(=O)Nc1ccc(c(c1)C(F)(F)F)[N+](=O)[O-]
IUPAC Name 2-methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide
Molecular Weight (g/mol) 276.21
Quantity 250 mg
Formula Weight 276.0722 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.