missing translation for 'onlineSavingsMsg'
Learn More

Fluvoxamine.maleate, Lipomed

Product Code. 30462588
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30462588

Brand: Lipomed LPMFLV1796MA10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Fluvoxamine.maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluvoxamine maleate
CAS 61718-82-9
Analyte or Component Names Fluvoxamine maleate
Shipping Condition Room Temperature
Molecular Formula C15 H21 F3 N2 O2 . C4 H4 O4
Synonym Fluvoxamine maleate, 2-[[[(1 E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethanamine (Z)-butenedioate, 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (1 E)-, (2 Z)-2-butenedioate (1:1), 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, (Z)-2-butenedioate (1:1), Avoksin, Avoxin, DU 23000, Depromel, Dumirox, Faverin, Faverin 50, Faverine, Fevarin, Floxyfral, Luvox, MK 264, Maveral, NSC 309469
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES COCCCC\C(=N/OCCN)\c1ccc(cc1)C(F)(F)F.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;2-[(E)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethanamine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 434.41
Quantity 10 mg
Formula Weight 434.1665 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.