Learn More
FMC63 scFv Mouse, FITC, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
Brand:  ACROBiosystems FM3-FY45-25tests
 View more versions of this product
Product Code. 16851936
  • 327.00 € / 25 tests
Description
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.
Specifications
Specifications
FMC63 scFv
Y45
Hybridoma
RUO
-20°C, aviod light
Monoclonal
FITC
25 Tests
Primary
IgG1