Learn More
FMC63 scFv Mouse, Unconjugated, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
Brand: ACROBiosystems FM3-Y45P1-250tests
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.Specifications
| FMC63 scFv | |
| Y45 | |
| Human (HEK293) | |
| RUO | |
| -20°C |
| Monoclonal | |
| Unconjugated | |
| 250 Tests | |
| Primary | |
| IgG1 |