missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMC63 scFv Mouse, Unconjugated, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
327.00 € - 7805.00 €
Specifications
| Antigen | FMC63 scFv |
|---|---|
| Clone | Y45 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Human (HEK293) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17176321
|
ACROBiosystems
FM3-Y45P1-25tests |
25 Tests |
327.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17166321
|
ACROBiosystems
FM3-Y45P1-250tests |
250 Tests |
2240.00 €
250 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17156321
|
ACROBiosystems
FM3-Y45P1-1000tests |
1000 Tests |
7805.00 €
1000 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.Specifications
| FMC63 scFv | |
| Monoclonal | |
| Human (HEK293) | |
| IgG1 | |
| -20°C |
| Y45 | |
| Unconjugated | |
| RUO | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title