missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMNL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24748
This item is not returnable.
View return policy
Description
FMNL2 Polyclonal specifically detects FMNL2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FMNL2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FHOD2, FLJ37546, formin homology 2 domain containing 2, Formin homology 2 domain-containing protein 2, formin-like 2, formin-like protein 2, KIAA1902 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 114793 | |
| Human, Mouse, Rat, Primate | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Q9Y613 | |
| FMNL2 | |
| A portion of amino acid 1-50 of human Formin-like protein 2 was used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, rat and mouse, 94% homologous in cow, 92% homologous in chicken and 88% homologous in opossum. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction