missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMNL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 351.00 €
Specifications
| Antigen | FMNL2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18187081
|
Novus Biologicals
NBP2-24748SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18787103
|
Novus Biologicals
NBP2-24748 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FMNL2 Polyclonal specifically detects FMNL2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FMNL2 | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| FHOD2, FLJ37546, formin homology 2 domain containing 2, Formin homology 2 domain-containing protein 2, formin-like 2, formin-like protein 2, KIAA1902 | |
| FMNL2 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, rat and mouse, 94% homologous in cow, 92% homologous in chicken and 88% homologous in opossum. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Primate | |
| Q9Y613 | |
| 114793 | |
| A portion of amino acid 1-50 of human Formin-like protein 2 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title