missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FMRP Monoclonal antibody specifically detects FMRP in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
Specifications
| Antigen | FMRP |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | S08-5G5 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA, Immunohistochemistry 1:20-1:500, Immunocytochemistry/ Immunofluorescence 1:100, Immunohistochemistry-Paraffin, Sandwich ELISA |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | FMRPFRAXAPOF, fragile X mental retardation 1, fragile X mental retardation 1 protein, MGC87458, POF1, premature ovarian failure 1, Protein FMR-1 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?