missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid, calcium salt pentahydrate, 95.0-105.0%

Product Code. 10751311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751311 1 g Glass bottle 1g
10411081 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751311 Supplier Thermo Scientific Acros Supplier No. 230310010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6035-45-6 | C20H21CaN7O7·5H2O | 601.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6035-45-6
Molecular Formula C20H21CaN7O7·5H2O
Molecular Weight (g/mol) 601.58
MDL Number MFCD00149465
InChI Key NPPBLUASYYNAIG-UHFFFAOYSA-L
Synonym folinic acid calcium pentahydrate
PubChem CID 131674093
IUPAC Name calcium;4-[[4-[(2-amino-5-formyl-4-oxido-7,8-dihydro-6H-pteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate;pentahydrate
SMILES C1C(N(C2=C(N1)N=C(N=C2[O-])N)C=O)CNC3=CC=C(C=C3)C(=O)NC(CCC(=O)[O-])C(=O)O.O.O.O.O.O.[Ca+2]

Specifications

Melting Point 240.0°C to 250.0°C
Color White-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4249
Specific Rotation + 14.30
Solubility Information Solubility in water: freely soluble in water. Other solubilities: practically insoluble in alcohol
Formula Weight 601.58
Percent Purity 95 to 105%
Physical Form Powder
Chemical Name or Material Folinic acid, calcium salt pentahydrate, 95.0-105.0%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.