accessibility menu, dialog, popup

UserName

CAS RN 6035-45-6

IUPAC Name: (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid calcium pentahydrate
Synonyms: folinic acid calcium pentahydrate
Molecular Weight (g/mol): 603.6
Molecular Formula: C20H33CaN7O12
InChi Key: DZNHRKNLTYYMQA-ZIGBGYJWSA-N
SMILES: O.O.O.O.O.[Ca].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)N2C=O)N1
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results