missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 1000 μg/mL in Water, Dr. Ehrenstorfer

Product Code. 30328218
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328218 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328218 Supplier Dr. Ehrenstorfer Supplier No. DREA13909000WA1000
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formaldehyde
CAS 50-00-0
Analyte or Component Names Formaldehyde
Shipping Condition -18°C
Molecular Formula C H2 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Methanal, BFV, F-gen, FM 282, FS 850A, Fannoform, Floguard 1015, Fordor, Formaldehyde-12C, Formalin, Formalin LM, Formalin Taisei, Formalith, Formic aldehyde, Formol, Fyde, Lysoform, Methaldehyde, Methyl aldehyde, Methylene oxide, Morbicid, NSC 298885, Optilyse, Oxomethane, Oxymethylene, Paraform, Superlysoform
InChI Formula InChI=1S/CH2O/c1-2/h1H2
SMILES C=O
IUPAC Name formaldehyde
Solvent or Matrix Water
Molecular Weight (g/mol) 30.03
Quantity 1 mL
Formula Weight 30.0106 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.