missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fosinopril Sodium, Mikromol™
Description
Fosinopril Sodium
Specifications
Specifications
| Chemical Name or Material | Fosinopril sodium |
| CAS | 88889-14-9 |
| Analyte or Component Names | Fosinopril sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H45 N O7 P . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | (4S)-4-Cyclohexyl-1-[[(R)-[(1S)-2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphinyl]acetyl]-L-proline, sodium salt, [1[S*(R*)],2alpha,4beta]-4-Cyclohexyl-1-[[[2-methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphinyl]acetyl]-L-proline, sodium salt, Acecor, Eosinopril, Fosinopril sodium, Monopril, SQ 28555, Secorvas, Staril |
| InChI Formula | InChI=1S/C30H46NO7P.Na/c1-4-28(33)37-30(22(2)3)38-39(36,18-12-11-15-23-13-7-5-8-14-23)21-27(32)31-20-25(19-26(31)29(34)35)24-16-9-6-10-17-24;/h5,7-8,13-14,22,24-26,30H,4,6,9-12,15-21H2,1-3H3,(H,34,35);/q;+1/p-1/t25-,26+,30-,39-;/m1./s1 |
| SMILES | [Na+].CCC(=O)O[C@H](O[P@](=O)(CCCCc1ccccc1)CC(=O)N2C[C@@H](C[C@H]2C(=O)[O-])C3CCCCC3)C(C)C |
| IUPAC Name | sodium;(2S,4S)-4-cyclohexyl-1-[2-[[(1R)-2-methyl-1-propanoyloxypropoxy]-(4-phenylbutyl)phosphoryl]acetyl]pyrrolidine-2-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?