missing translation for 'onlineSavingsMsg'
Learn More

FOXO3 Antibody (CL13603) [DyLight 680], Novus Biologicals™

Product Code. 30524481 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30524481 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30524481 Supplier Novus Biologicals Supplier No. NBP343834FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXO3 Monoclonal antibody specifically detects FOXO3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXO3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13603
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of O43524, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 2309
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.